Få mere at vide
2-Chloronaphthalene-d7, CDN
Artikelnummer. 30321914
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321914 0.05 g
30320834 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321914 Leverandør CDN Leverandørnr. CDND24620.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloronaphthalene-d7
CAS 93951-84-9
Molekylær formel C10 2H7 Cl
Mængde 0.05 g
Synonym Naphthalene-1,2,3,4,5,6,8-d7, 7-chloro- (9 CI), 2-Chloronaphthalene D7, 2-Chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene, 2-Chloronaphthalene-1,3,4,5,6,7,8-D7
InChI formel InChI=1 S/C10H7Cl/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c2c([2 H])c(Cl)c([2 H])c([2 H])c2c1[2 H]
IUPAC navn 2-chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene
Molekylvægt (g/mol) 169.66
Formel vægt 169.0676 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity