missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7021674
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3747-74-8
C10H9Cl2N
214.089
MFCD00012734
WDETYCRYUBGKCE-UHFFFAOYSA-N
2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
3083823
2-(chloromethyl)quinoline;hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl
This item is not returnable. View return policy
3747-74-8
C10H9Cl2N
214.089
MFCD00012734
WDETYCRYUBGKCE-UHFFFAOYSA-N
2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
3083823
2-(chloromethyl)quinoline;hydrochloride
C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 184°C to 190°C
Quantity 1 g
Beilstein 3565832
Formula Weight 214.1
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)quinoline hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'