missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Chloromethyl)benzothiazole, 95%

Brand:  Alfa Aesar™ H64781.MD

Additional Details : CAS Number : 37859-43-1 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15492129

  • 313.98 DKK / 250mg


Estimated Shipment
27-10-2020
Add to basket

Specifications

Chemical Identifiers

37859-43-1
183.653
SERUZNHRWBXDOX-UHFFFAOYSA-N
304978
C1=CC=C2C(=C1)N=C(S2)CCl
C8H6ClNS
MFCD05664944
2-chloromethyl-1,3-benzothiazole, 2-chloromethyl benzothiazole, 2-chloromethyl benzo d thiazole, benzothiazole, 2-chloromethyl, 2-chloromethylbenzothiazole, 2-chloromethyl-benzothiazole, 2-chloromethylbenzthiazole, acmc-1ag8v, ksc776s1l, 2-chloromethylbenzo d thiazole
2-(chloromethyl)-1,3-benzothiazole

Specifications

2-(Chloromethyl)benzothiazole
88°C to 99°C (0.3mmHg)
250mg
C8H6ClNS
120805
SERUZNHRWBXDOX-UHFFFAOYSA-N
2-(chloromethyl)-1,3-benzothiazole
304978
Solid
Moisture sensitive
34°C
37859-43-1
MFCD05664944
2-chloromethyl-1,3-benzothiazole, 2-chloromethyl benzothiazole, 2-chloromethyl benzo d thiazole, benzothiazole, 2-chloromethyl, 2-chloromethylbenzothiazole, 2-chloromethyl-benzothiazole, 2-chloromethylbenzthiazole, acmc-1ag8v, ksc776s1l, 2-chloromethylbenzo d thiazole
C1=CC=C2C(=C1)N=C(S2)CCl
183.653
183.65
95%
Videos
SDS Documents