missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)benzimidazole, 95%, Thermo Scientific Chemicals

Catalog Number p-7028308
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4857-04-9
This item is not returnable. View return policy
4857-04-9

2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 141°C to 143°C (decomposition)
Molecular Formula C8H7ClN2
MDL Number MFCD00005602
Quantity 5 g
Synonym 2-chloromethylbenzimidazole, 2-chloromethyl benzimidazole, 2-chloromethyl-1h-benzo d imidazole, 2-chloromethyl-1h-benzimidazole, 2-chloromethyl-1h-1,3-benzodiazole, 2-chloromethyl-1h-benzoimidazole, 1h-benzimidazole, 2-chloromethyl, dimezole-7, 2-chloromethyl-1h-1,3-benzimidazole
Solubility Information Insoluble in water.
InChI Key SPMLMLQATWNZEE-UHFFFAOYSA-N
SMILES ClCC1=NC2=CC=CC=C2N1
IUPAC Name 2-(chloromethyl)-1H-benzimidazole
Molecular Weight (g/mol) 166.61
PubChem CID 78571
Formula Weight 166.61
Percent Purity 96%
Chemical Name or Material 2-(Chloromethyl)benzimidazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'