missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloromethyl-4-nitrophenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 2973-19-5 |
Melting Point | 128.0°C to 132.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass Bottle |
Molecular Formula | C7H6ClNO3 |
Linear Formula | ClCH2C6H3(NO2)OH |
Quantity | 1 g |
Beilstein | 06,367 |
Synonym | 2-chloromethyl-4-nitrophenol, phenol, 2-chloromethyl-4-nitro, alpha-chloro-4-nitro-o-cresol, 2-hydroxy-5-nitrobenzyl chloride, 2-hidroxi-5-nitrobenzilclorura romanian, 2-chloromethyl-4-nitro-phenol, phenol,2-chloromethyl-4-nitro, 2-hidroxi-5-nitrobenzilclorura, acmc-1ccok, 4-nitro-2-chloromethylphenol |
Show More |
Safety and Handling
Product Identifier
- 2-Chloromethyl-4-nitrophenol
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'