Få mere at vide
2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride, Mikromol™
Artikelnummer. 30463322
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463322 4 x 25 mg
30050162 25 mg
18042070 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463322 Leverandør Mikromol Leverandørnr. MM0618.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
CAS 153259-31-5
Analyt- eller komponentnavne 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C11 H16 Cl N O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym 2-Chloromethyl-3-methyl-4-(3-methoxypropoxy)pyridine Hydrochloride
InChI formel InChI=1S/C11H16ClNO2.ClH/c1-9-10(8-12)13-5-4-11(9)15-7-3-6-14-2;/h4-5H,3,6-8H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.COCCCOc1ccnc(CCl)c1C
IUPAC navn 2-(chloromethyl)-4-(3-methoxypropoxy)-3-methylpyridine;hydrochloride
Molekylvægt (g/mol) 266.16
Mængde 4 x 25 mg
Formel vægt 265.0636 g/mol