missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Chloromethyl-3,5-dimethylpyridin-4-ol, TRC

Artikelnummer. 17974525
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974525 250 mg
17933485 1 g
17924915 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17974525 Leverandør Toronto Research Chemicals Leverandørnr. TRCC369200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220771-03-9
Molekylær formel C8 H10 Cl N O
Molekylvægt (g/mol) 171.6241
Synonym 4-Pyridinol, 2-(chloromethyl)-3,5-dimethyl-, 2-(Chloromethyl)-3,5-dimethyl-4-pyridinol, 2-Chloromethyl-3,5-dimethyl-4-hydroxypyridine, 2-(Chloromethyl)-3,5-dimethyl-4-pyridinol, Pyridoxine Impurity
IUPAC navn 2-(chloromethyl)-3,5-dimethylpyridin-4-ol
SMIL Cc1cnc(CCl)c(C)c1O

Specifications

Kemisk navn eller materiale 2-Chloromethyl-3,5-dimethylpyridin-4-ol
Mængde 250 mg
InChI formel InChI=1S/C8H10ClNO/c1-5-4-10-7(3-9)6(2)8(5)11/h4H,3H2,1-2H3,(H,10,11)
Formel vægt 171.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.