Få mere at vide
2-Chloromethyl-3,5-dimethylpyridin-4-ol, TRC
Artikelnummer. 17933485
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17933485 1 g
17974525 250 mg
17924915 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17933485 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3692001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 220771-03-9
Molekylær formel C8 H10 Cl N O
Molekylvægt (g/mol) 171.6241
Synonym 4-Pyridinol, 2-(chloromethyl)-3,5-dimethyl-, 2-(Chloromethyl)-3,5-dimethyl-4-pyridinol, 2-Chloromethyl-3,5-dimethyl-4-hydroxypyridine, 2-(Chloromethyl)-3,5-dimethyl-4-pyridinol, Pyridoxine Impurity
IUPAC navn 2-(chloromethyl)-3,5-dimethylpyridin-4-ol
SMIL Cc1cnc(CCl)c(C)c1O
Tekniske data
Kemisk navn eller materiale 2-Chloromethyl-3,5-dimethylpyridin-4-ol
Mængde 1 g
InChI formel InChI=1S/C8H10ClNO/c1-5-4-10-7(3-9)6(2)8(5)11/h4H,3H2,1-2H3,(H,10,11)
Formel vægt 171.0451