missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature.
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature.
Specifications
Melting Point | ∽212°C (decomposition) |
Quantity | 10 g |
Sensitivity | Air sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 179.61 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloroindole-3-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'