missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals

Catalog Number p-7027376
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
627-42-9
C3H7ClO
94.54
MFCD00000972
XTIGGAHUZJWQMD-UHFFFAOYSA-N
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
12316
1-chloro-2-methoxyethane
COCCCl
This item is not returnable. View return policy
627-42-9
C3H7ClO
94.54
MFCD00000972
XTIGGAHUZJWQMD-UHFFFAOYSA-N
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
12316
1-chloro-2-methoxyethane
COCCCl

2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

Solubility
Soluble in water 60 g/L at 20°C.

Notes
Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Density 1.04
Boiling Point 89°C to 90°C
Flash Point 8°C (46°F)
Linear Formula CH3OCH2CH2Cl
Refractive Index 1.409
Quantity 25 g
UN Number UN3271
Solubility Information Soluble in water 60g/L at 20°C.
Formula Weight 94.54
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl methyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'