Learn More
2-Chloroethyl ethyl ether, 98+%, Thermo Scientific Chemicals
Description
It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.
Solubility
Miscible with water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | -60°C |
Boiling Point | 107°C to 108°C |
Flash Point | 6°C (43°F) |
Quantity | 25 g |
UN Number | UN3271 |
Formula Weight | 108.57 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Chloroethyl ethyl ether |
RUO – Research Use Only