missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Chloroethyl Chloroformate, TRC

Artikelnummer. 30350813
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350813 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30350813 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3672451G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Chloroethyl Chloroformate
CAS 627-11-2
Molekylær formel C3H4Cl2O2
Mængde 1 g
Synonym beta-Chloroethyl chloroformate, Chloroformic Acid 2-Chloroethyl Ester, (2-Chloroethoxy)carbonyl Chloride, 2-Chloroethyl Carbonochloridate, 2-Chloroethyl Chlorocarbonate, 2-Chloroethyl Chloroformate, Chloroethyl Chloroformate, Chloroformic Acid 2-Chloroethyl Ester
InChI formel InChI=1S/C3H4Cl2O2/c4-1-2-7-3(5)6/h1-2H2
SMIL ClCCOC(=O)Cl
IUPAC navn 2-chloroethyl carbonochloridate
Molekylvægt (g/mol) 142.97
Formel vægt 141.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.