missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanol, 99%, Alfa Aesar™

Brand:  Alfa Aesar 036636.K2

Additional Details : CAS Number : 107-07-3 Weight : 1.00000kg Transport : UN number : 1135 Chem class : 6.1 Pack group : I

Quantity 1L

 View more versions of this product

Product Code. 15401467

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance
View alternative products »


Specifications

Chemical Identifiers

107-07-3
80.511
SZIFAVKTNFCBPC-UHFFFAOYSA-N
34
2-chloroethanol
C2H5ClO
MFCD00002829
Ethylene chlorohydrin
CHEBI:28200
C(CCl)O

Specifications

2-Chloroethanol
55° (131°F)
129°C
-63°C
107-07-3
99%
ClCH2CH2OH
UN1135
14,3793
SZIFAVKTNFCBPC-UHFFFAOYSA-N
2-chloroethanol
34
80.51
1.202g/mL
Ether-like
Yellow
1L
1.4410
C2H5ClO
MFCD00002829
878139
Ethylene chlorohydrin
C(CCl)O
80.511
CHEBI:28200
Liquid
Videos
SDS