Få mere at vide
(±)-2-Chlorobutane-d9, CDN
Artikelnummer. 30316245
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316245 0.05 g -
30311088 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316245 Leverandør CDN Leverandørnr. CDND66830.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chlorobutane-D9
CAS 175540-77-9
Molekylær formel C4 D9 Cl
Mængde 0.05 g
Synonym Butane-1,1,1,2,2,3,4,4,4-d9, 3-chloro- (9 CI), (±)-2-Chlorobutane-d9, 3-Chlorobutane-1,1,1,2,2,3,4,4,4-d9, 2-Chlorobutane-D9
InChI formel InChI=1 S/C4H9Cl/c1-3-4(2)5/h4H,3H2,1-2H3/i1D3,2D3,3D2,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(Cl)C([2 H])([2 H])[2 H]
IUPAC navn 2-chloro-1,1,1,2,3,3,4,4,4-nonadeuteriobutane
Molekylvægt (g/mol) 101.6227
Formel vægt 101.0958 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity