missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobiphenyl, TRC

Product Code. 30366267
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30366267 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366267 Supplier Toronto Research Chemicals Supplier No. TRCC364793100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale PCB No. 1
CAS 2051-60-7
Molekylær formel C12 H9 Cl
Mængde 100 mg
Synonym 1,1'-Biphenyl, 2-chloro-, Biphenyl, 2-chloro- (6CI,7CI,8CI), 2-Chloro-1,1'-biphenyl, 1-Chloro-2-phenylbenzene, 2-Chlorobiphenyl, 2-Chlorodiphenyl, 2-Monochlorobiphenyl, NSC 67354, PCB 1, o-Chlorobiphenyl, 2-MonoCB
InChI formel InChI=1S/C12H9Cl/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H
SMIL Clc1ccccc1c2ccccc2
IUPAC navn 1-chloro-2-phenylbenzene
Molekylvægt (g/mol) 188.65
Formel vægt 188.0393
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.