missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzophenone, 99+%, Thermo Scientific Chemicals

Catalog Number p-7022894
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
5162-03-8
C13H9ClO
216.66
MFCD00000558
VMHYWKBKHMYRNF-UHFFFAOYSA-N
This item is not returnable. View return policy
5162-03-8
C13H9ClO
216.66
MFCD00000558
VMHYWKBKHMYRNF-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 47°C
Boiling Point 330°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1869594
Synonym 2-chlorobenzophenone, 2-chlorophenyl phenyl methanone, methanone, 2-chlorophenyl phenyl, o-chlorobenzophenone, 2-chlorophenyl phenylmethanone, benzophenone, 2-chloro, 2-chlorophenyl-phenylmethanone, 2-chlorobenyl phenyl ketone, 2-chloro-phenyl-phenyl-methanone, benzophenone, 2-chloro-6ci,7ci,8ci
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
IUPAC Name (2-chlorophenyl)-phenylmethanone
PubChem CID 78838
Formula Weight 216.67
Percent Purity ≥99%
Chemical Name or Material 2-Chlorobenzophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'