Få mere at vide
2-Chlorobenzoic Acid-d4, TRC
Artikelnummer. 30358418
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30358418 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358418 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3646431G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chlorobenzoic Acid-d4
CAS 1219795-28-4
Molekylær formel C7 D4 H Cl O2
Mængde 1 g
Synonym o-Chlorobenzoic Acid-d4, 2-CBA-d4, NSC 15042-d4, NSC 32737-d4, o-Chlorobenzoic Acid-d4
InChI formel InChI=1S/C7H5ClO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(Cl)c1[2H]
IUPAC navn 2-chloro-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 160.59
Formel vægt 160.0229