Få mere at vide
2-Chloroaniline-4,6-d2, TRC
Artikelnummer. 30366180
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30366180 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366180 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36437110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloroaniline-4,6-d2
CAS 347840-10-2
Molekylær formel C6 D2 H4 Cl N
Mængde 10 mg
Synonym Benzen-2,4-d2-amine, 6-chloro- (9CI), 2-Chloroaniline-4,6-d2, 2-Chloroaniline-4,6-D2, 2-Chloroaniline-d2, 6-Chlorobenzen-2,4-d2-amine, 2-Chlorobenzen-4,6-d2-amine
InChI formel InChI=1S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2/i1D,4D
SMIL [2H]c1cc([2H])c(N)c(Cl)c1
IUPAC navn 2-chloro-4,6-dideuterioaniline
Molekylvægt (g/mol) 129.584
Formel vægt 129.031