Få mere at vide
2-Chloroaniline-4,6-d2, CDN
Artikelnummer. 30320338
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320338 1 g
30312582 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320338 Leverandør CDN Leverandørnr. CDND37001G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloroaniline-4,6-d2
CAS 347840-10-2
Molekylær formel C6 D2 H4 Cl N
Mængde 1 g
Synonym Benzen-2,4-d2-amine, 6-chloro- (9 CI), 2-Chloroaniline-4,6-d2, 2-Chloroaniline-4,6-D2, 2-Chloroaniline-d2, 6-Chlorobenzen-2,4-d2-amine, 2-Chlorobenzen-4,6-d2-amine
InChI formel InChI=1 S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4 H,8H2/i1D,4 D
SMIL [2 H]c1cc([2 H])c(N)c(Cl)c1
IUPAC navn 2-chloro-4,6-dideuterioaniline
Molekylvægt (g/mol) 129.584
Formel vægt 129.031 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity