Learn More
2-Chloroacetamide, Sigma-Aldrich™
Product Code. 15898750
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898750 100 g 100g
15808760 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15898750 Supplier Sigma Aldrich Supplier No. 8024120100
 Restricted Item
Forventet forsendelse: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-07-2
Grade Research
Molecular Weight (g/mol) 93.51
Specifications
Melting Point 115°C to 118°C
MDL Number MFCD00008027
Quantity 100 g
Synonym 2-Chloroacetamide, 2-Chloroacetamide