Få mere at vide
2-Chloroacetamide-d4, TRC
Artikelnummer. 30353726
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30353726 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353726 Leverandør Toronto Research Chemicals Leverandørnr. TRCC353502100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloroacetamide N,N,2,2-d4
CAS 122775-20-6
Molekylær formel C2 2H4 Cl N O
Mængde 100 mg
Synonym Acetamide-N,N,2,2-d4, 2-chloro- (9CI), 2-Chloro-N,N,2,2-tetradeuterioacetamide, 2-Chloroacetamide-d4, 2-Chloroacetamide-D4, 2-Chloroacetamide-N,N,2,2-d4
InChI formel InChI=1S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)/i1D2/hD2
SMIL [2H]N([2H])C(=O)C([2H])([2H])Cl
IUPAC navn 2-chloro-N,N,2,2-tetradeuterioacetamide
Molekylvægt (g/mol) 97.54
Formel vægt 97.0232