Få mere at vide
2-Chloro-N,N-dimethylethylamine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Chloro-N,N-dimethylethylamine Hydrochloride |
| CAS | 4584-46-7 |
| Molekylær formel | C4H11Cl2N |
| Mængde | 100 g |
| Synonym | (2-Chloroethyl)dimethylamine Hydrochloride, (β-Chloroethyl)dimethylamine Hydrochloride, 1-(Dimethylamino)-2-chloroethane-d6 Hydrochloride, 1-Chloro-2-(dimethylamino)ethane Hydrochloride, 1-Chloro-2-N,N-dimethylaminoethane Hydrochloride, 1-Chloro-2-dimethylaminoethane Monohydrochloride, 2-(Dimethylamino)chloroethane Hydrochloride, 2-(Dimethylamino)ethyl Chloride Hydrochloride, 2-(N,N-Dimethylamino)ethyl Chloride Hydrochloride, 2-Chloro-N,N-dimethylethanamine Hydrochloride, 2-Chloro-N,N-dimethylethaneamine Hydrochloride, 2-Chloro-N,N-dimethylethylamine Hydrochloride, 2-Chloroethyl-N,N-dimethylamine Hydrochloride, 2-Chloroethyldimethylamine Monohydrochloride, 2-Chloroethyldimethylammonium Chloride, Chloro(dimethylamino)ethane Hydrochloride, Chloroethyldimethylamine Hydrochloride, Dimethylaminoethyl Chloride Hydrochloride, N,N-Dimethyl-2-chloroethanamine Hydrochloride, N,N-Dimethyl-2-chloroethylamine Hydrochloride, N,N-Dimethyl-N-(2-chloroethyl)amine Hydrochloride, N-(2-Chloroethyl)-N,N-dimethylamine Hydrochloride, N-(2-Chloroethyl)-N,N-dimethylammonium Chloride, N-(2-Chloroethyl)dimethylamine Hydrochloride, β-Chloroethyl-N,N-dimethylamine Hydrochloride, β-Dimethylaminoethyl Chloride Hydrochloride |
| InChI formel | InChI=1S/C4H10ClN.ClH/c1-6(2)4-3-5;/h3-4H2,1-2H3;1H |
| SMIL | Cl.CN(C)CCCl |
| IUPAC navn | 2-chloro-N,N-dimethylethanamine;hydrochloride |
| Molekylvægt (g/mol) | 144.04 |
| Formel vægt | 143.0269 |