Få mere at vide
2-Chloro-N,N-dimethyl-d6-ethylamine HCl, CDN
Artikelnummer. 30324032
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324032 0.5 g 0.5g
30310065 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324032 Leverandør CDN Leverandørnr. CDND65260.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro-N,N-dimethylethylamine dimethyl-d6 hydrochloride
CAS 97941-91-8
Molekylær formel C4 2H6 H4 Cl N . Cl H
Mængde 0.5 g
InChI formel InChI=1 S/C4H10ClN.ClH/c1-6(2)4-3-5;/h3-4H2,1-2H3;1 H/i1D3,2D3;
SMIL Cl.[2 H]C([2 H])([2 H])N(CCCl)C([2 H])([2 H])[2 H]
IUPAC navn 2-chloro-N,N-bis(trideuteriomethyl)ethanamine;hydrochloride
Molekylvægt (g/mol) 150.08
Formel vægt 149.0645 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity