missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-diethyl-4-nitroaniline, 96%, Thermo Scientific™

Catalog Number p-7054847
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
86-49-7
C10H13ClN2O2
228.676
PCVJAZCSKCCKPQ-UHFFFAOYSA-N
benzenamine, 2-chloro-n,n-diethyl-4-nitro, aniline, 2-chloro-n,n-diethyl-4-nitro, n,n-diethyl-2-chloro-4-nitroaniline, 2-chloro-4-nitro-n,n-diethylaniline, 2-chloro-n, n-diethyl-4-nitroaniline, n-2-chloro-4-nitrophenyl-n,n-diethylamine #
66580
2-chloro-N,N-diethyl-4-nitroaniline
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])Cl
This item is not returnable. View return policy
86-49-7
C10H13ClN2O2
228.676
PCVJAZCSKCCKPQ-UHFFFAOYSA-N
benzenamine, 2-chloro-n,n-diethyl-4-nitro, aniline, 2-chloro-n,n-diethyl-4-nitro, n,n-diethyl-2-chloro-4-nitroaniline, 2-chloro-4-nitro-n,n-diethylaniline, 2-chloro-n, n-diethyl-4-nitroaniline, n-2-chloro-4-nitrophenyl-n,n-diethylamine #
66580
2-chloro-N,N-diethyl-4-nitroaniline
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Formula Weight 228.68
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Chloro-N,N-diethyl-4-nitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'