missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(hydroxymethyl)acetamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081992
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
2832-19-1
C3H6ClNO2
123.54
MFCD00021961
TXNSZCSYBXHETP-UHFFFAOYSA-N
2-chloro-n-hydroxymethyl acetamide, n-hydroxymethyl-2-chloroacetamide, acetamide, 2-chloro-n-hydroxymethyl, n-methylolchloroacetamide, grota, chloracetamide-n-metholol, n-methylol-chloracetamide, grotan df-35, caswell no. 192bb, chloracetamide-n-methanol
62466
2-chloro-N-(hydroxymethyl)acetamide
OCNC(=O)CCl
This item is not returnable. View return policy
2832-19-1
C3H6ClNO2
123.54
MFCD00021961
TXNSZCSYBXHETP-UHFFFAOYSA-N
2-chloro-n-hydroxymethyl acetamide, n-hydroxymethyl-2-chloroacetamide, acetamide, 2-chloro-n-hydroxymethyl, n-methylolchloroacetamide, grota, chloracetamide-n-metholol, n-methylol-chloracetamide, grotan df-35, caswell no. 192bb, chloracetamide-n-methanol
62466
2-chloro-N-(hydroxymethyl)acetamide
OCNC(=O)CCl

An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.

Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 97°C to 103°C
Quantity 10 g
UN Number UN3261
Beilstein 1811765
Sensitivity Hygroscopic
Solubility Information Very soluble in water. Poorly soluble in aliphatic hydrocarbons.
Formula Weight 123.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-N- (hydroxymethyl)acetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'