missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog Number 15455086
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
331845-70-6
C14H17ClN2O3
296.75
MFCD01115617
FSSFCAMVJVCOMW-UHFFFAOYSA-N
2-chloro-n-cyclohexyl-4-nitro-n-methylbenzamide, 2-chloro-n-cyclohexyl-n-methyl-4-nitro-benzamide, 2-chloro-4-nitrophenyl-n-cyclohexyl-n-methylcarboxamide
799284
2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide
CN(C1CCCCC1)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
331845-70-6
C14H17ClN2O3
296.75
MFCD01115617
FSSFCAMVJVCOMW-UHFFFAOYSA-N
2-chloro-n-cyclohexyl-4-nitro-n-methylbenzamide, 2-chloro-n-cyclohexyl-n-methyl-4-nitro-benzamide, 2-chloro-4-nitrophenyl-n-cyclohexyl-n-methylcarboxamide
799284
2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide
CN(C1CCCCC1)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 296.75
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'