missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide, 97%
Brand: Alfa Aesar™ H59539.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
331845-70-6 | |
296.751 | |
FSSFCAMVJVCOMW-UHFFFAOYSA-N | |
799284 | |
CN(C1CCCCC1)C(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl |
C14H17ClN2O3 | |
MFCD01115617 | |
2-chloro-n-cyclohexyl-4-nitro-n-methylbenzamide, 2-chloro-n-cyclohexyl-n-methyl-4-nitro-benzamide, 2-chloro-4-nitrophenyl-n-cyclohexyl-n-methylcarboxamide | |
2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide |
Specifications
2-Chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide | |
331845-70-6 | |
MFCD01115617 | |
FSSFCAMVJVCOMW-UHFFFAOYSA-N | |
2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide | |
799284 | |
97% |
1g | |
C14H17ClN2O3 | |
2-chloro-n-cyclohexyl-4-nitro-n-methylbenzamide, 2-chloro-n-cyclohexyl-n-methyl-4-nitro-benzamide, 2-chloro-4-nitrophenyl-n-cyclohexyl-n-methylcarboxamide | |
CN(C1CCCCC1)C(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl | |
296.751 | |
296.75 |