Få mere at vide
2-Chloro-N-(2-chloro-4-methyl-3-pyridinyl)-3-pyridinecarboxamide, Mikromol™
Artikelnummer. 30052686
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052686 25 mg
30051351 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052686 Leverandør Mikromol Leverandørnr. MM1146.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloro-N-(2-chloro-4-methyl-3-pyridinyl)-3-pyridinecarboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro-N-(2-chloro-4-methyl-3-pyridinyl)-3-pyridinecarboxamide
CAS 133627-46-0
Analyt- eller komponentnavne 2-Chloro-N-(2-chloro-4-methyl-3-pyridinyl)-3-pyridinecarboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C12 H9 Cl2 N3 O
Certificeringer/compliance ISO 17025
Synonym 2-Chloro-N-(2-chloro-4-methyl-3-pyridinyl)-3-pyridinecarboxamide, 2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)-3-pyridine carboxamide, Picnic
InChI formel InChI=1S/C12H9Cl2N3O/c1-7-4-6-16-11(14)9(7)17-12(18)8-3-2-5-15-10(8)13/h2-6H,1H3,(H,17,18)
SMIL Cc1ccnc(Cl)c1NC(=O)c2cccnc2Cl
IUPAC navn 2-chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide
Molekylvægt (g/mol) 282.13
Mængde 25 mg
Formel vægt 281.0123 g/mol