Få mere at vide
2-Chloro Hippuric Acid-d3, TRC
Artikelnummer. 30366734
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30366734 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366734 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36654725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro Hippuric Acid-d3
CAS 1794816-82-2
Molekylær formel C9H5D3ClNO3
Mængde 25 mg
Synonym (2-Chlorobenzoylamino)acetic Acid-d3, N-(2-Chlorobenzoyl)glycine-d3, N-(o-Chlorobenzoyl)glycine-d3, o-Chlorobenzoylglycine-d3, o-Chlorohippuric Acid-d3
InChI formel InChI=1S/C9H8ClNO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)/i3D,5D2
SMIL O=C(NC([2H])([2H])C(O)=O)C1=C([2H])C=CC=C1Cl
IUPAC navn 2-[(2-chloro-6-deuteriobenzoyl)amino]-2,2-dideuterioacetic acid
Molekylvægt (g/mol) 216.64