Få mere at vide
2-Chloro-ethanamine-d4 Hydrochloride, TRC
Artikelnummer. 30351274
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30351274 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351274 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36573710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro-ethanamine 1,1,2,2-d4 hydrochloride
CAS 172333-26-5
Molekylær formel C2 2H4 H2 Cl N . Cl H
Mængde 10 mg
Synonym 2-Chloroethan-1,1,2,2-d4-amine hydrochloride (9CI)
InChI formel InChI=1S/C2H6ClN.ClH/c3-1-2-4;/h1-2,4H2;1H/i1D2,2D2;
SMIL Cl.[2H]C([2H])(N)C([2H])([2H])Cl
IUPAC navn 2-chloro-1,1,2,2-tetradeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 120.01
Formel vægt 119.0207