Få mere at vide
2-Chloro-8-methoxy-dibenz[b,f][1,4]oxazepin-11(10H)-one, TRC
Artikelnummer. 30359712
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359712 1 g
30350814 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359712 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3679851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797131-66-8
Molekylær formel C14H10ClNO3
Molekylvægt (g/mol) 275.69
Synonym Dibenz[b,f][1,4]oxazepin-11(10H)-one, 2-chloro-8-methoxy- (ACI), 2-Chloro-8-methoxydibenz[b,f][1,4]oxazepin-11(10H)-one (ACI)
IUPAC navn 8-chloro-3-methoxy-5H-benzo[b][1,4]benzoxazepin-6-one
SMIL COc1ccc2Oc3ccc(Cl)cc3C(=O)Nc2c1
Tekniske data
Kemisk navn eller materiale 2-Chloro-8-methoxy-dibenz[b,f][1,4]oxazepin-11(10H)-one
Mængde 1 g
InChI formel InChI=1S/C14H10ClNO3/c1-18-9-3-5-13-11(7-9)16-14(17)10-6-8(15)2-4-12(10)19-13/h2-7H,1H3,(H,16,17)
Formel vægt 275.0349