Få mere at vide
2-Chloro-6-(trifluoromethoxy)benzo[d]thiazole, TRC
Artikelnummer. 30364003
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364003 250 mg
30354088 50 mg
30359198 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364003 Leverandør Toronto Research Chemicals Leverandørnr. TRCC597533250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 133840-96-7
Molekylær formel C8H3NOF3SCl
Molekylvægt (g/mol) 253.62
IUPAC navn 2-chloro-6-(trifluoromethoxy)-1,3-benzothiazole
SMIL FC(F)(F)Oc1ccc2nc(Cl)sc2c1
Tekniske data
Kemisk navn eller materiale 2-Chloro-6-(trifluoromethoxy)benzo[d]thiazole
Mængde 250 mg
InChI formel InChI=1S/C8H3ClF3NOS/c9-7-13-5-2-1-4(3-6(5)15-7)14-8(10,11)12/h1-3H
Formel vægt 252.9576