Learn More
2-Chloro-6-methyl-4-nitroaniline, 98%, Thermo Scientific™
Catalog Number 11494837
Click to view available options
Quantity:
1 g
5 g
69951-02-6
C7H7ClN2O2
186.595
MFCD01313703
HVPVUAIKGOKEEE-UHFFFAOYSA-N
2733460
2-chloro-6-methyl-4-nitroaniline
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-]
69951-02-6
C7H7ClN2O2
186.595
MFCD01313703
HVPVUAIKGOKEEE-UHFFFAOYSA-N
2733460
2-chloro-6-methyl-4-nitroaniline
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-]
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 174°C to 176°C
Quantity 1 g
Synonym 2-chloro-4-nitro-6-methylaniline, 2-chloro-6-methyl-4-nitro-phenylamine, benzenamine,2-chloro-6-methyl-4-nitro, benzenamine, 2-chloro-6-methyl-4-nitro, 6-chloro-2-methyl-4-nitrophenylamine, pubchem4373, chloromethylnitroaniline, acmc-1b462, 6-chloro-2-methyl-4-nitroaniline, 2-chloro-6-methyl-4-nitrophenylamine
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methyl-4-nitroaniline