Få mere at vide
2-Chloro-6-(2-hydroxybenzylamino)-9-isopropylpurine, TRC
Artikelnummer. 30366134
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366134 50 mg
30351684 10 mg
30361503 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366134 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36661050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500568-72-9
Molekylær formel C15 H16 Cl N5 O
Molekylvægt (g/mol) 317.77
IUPAC navn 2-[[(2-chloro-9-propan-2-ylpurin-6-yl)amino]methyl]phenol
SMIL CC(C)n1cnc2c(NCc3ccccc3O)nc(Cl)nc12
Tekniske data
Kemisk navn eller materiale 2-Chloro-6-(2-hydroxybenzylamino)-9-isopropylpurine
Mængde 50 mg
InChI formel InChI=1S/C15H16ClN5O/c1-9(2)21-8-18-12-13(19-15(16)20-14(12)21)17-7-10-5-3-4-6-11(10)22/h3-6,8-9,22H,7H2,1-2H3,(H,17,19,20)
Formel vægt 317.1043