missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-picoline, 94%, Thermo Scientific™

Catalog Number p-2698091
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18368-64-4
C6H6ClN
127.57
MFCD00792460
VXLYOURCUVQYLN-UHFFFAOYSA-N
2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine
581393
2-chloro-5-methylpyridine
CC1=CC=C(Cl)N=C1
This item is not returnable. View return policy
18368-64-4
C6H6ClN
127.57
MFCD00792460
VXLYOURCUVQYLN-UHFFFAOYSA-N
2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine
581393
2-chloro-5-methylpyridine
CC1=CC=C(Cl)N=C1
CAS Min % 92.0
CAS Max % 100.0
Color Orange-Pink or Straw-Yellow to Yellow
Density 1.1690g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.53
Quantity 5 g
Specific Gravity 1.169
Formula Weight 127.57
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-chloro-5-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'