missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(pentafluorothio)benzaldehyde, 97%, Thermo Scientific™

Catalog Number 15467487
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1431329-76-8
C7H4ClF5OS
266.61
MFCD22988924
SWBTYNKTVFWDHW-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzaldehyde, 2-chloro-5-pentafluorosulfur benzaldehyde, 2-chloro-5-pentafluoro-??-sulfanyl benzaldehyde
73995187
2-chloro-5-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C=O)Cl
This item is not returnable. View return policy
1431329-76-8
C7H4ClF5OS
266.61
MFCD22988924
SWBTYNKTVFWDHW-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzaldehyde, 2-chloro-5-pentafluorosulfur benzaldehyde, 2-chloro-5-pentafluoro-??-sulfanyl benzaldehyde
73995187
2-chloro-5-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 266.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(pentafluorothio)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'