missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitronicotinic acid, 95%, Thermo Scientific™

Catalog Number p-7057346
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42959-38-6
C6H3ClN2O4
202.55
MFCD06658402
WOFZRBCITDVRON-UHFFFAOYSA-N
2-chloro-5-nitronicotinic acid, 2-chloro-5-nitropyridine-3-carboxylic acid, 2-chloro-5-nitro-nicotinic acid, 3-carboxy-2-chloro-5-nitropyridine, 3-carboxy-2-chloro-5-nitro pyridine, 2-chloro-5-nitronicotinicacid, pubchem9631, acmc-209js2, ksc235o0f, chloro-5-nitro nicotinic acid
12542664
2-chloro-5-nitropyridine-3-carboxylic acid
OC(=O)C1=CC(=CN=C1Cl)[N+]([O-])=O
This item is not returnable. View return policy
42959-38-6
C6H3ClN2O4
202.55
MFCD06658402
WOFZRBCITDVRON-UHFFFAOYSA-N
2-chloro-5-nitronicotinic acid, 2-chloro-5-nitropyridine-3-carboxylic acid, 2-chloro-5-nitro-nicotinic acid, 3-carboxy-2-chloro-5-nitropyridine, 3-carboxy-2-chloro-5-nitro pyridine, 2-chloro-5-nitronicotinicacid, pubchem9631, acmc-209js2, ksc235o0f, chloro-5-nitro nicotinic acid
12542664
2-chloro-5-nitropyridine-3-carboxylic acid
OC(=O)C1=CC(=CN=C1Cl)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 140°C to 143°C
Color White to Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 202.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-nitronicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'