missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-7027495
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16588-02-6
C7H3ClN2O2
182.563
MFCD00007292
ZGILLTVEEBNDOB-UHFFFAOYSA-N
benzonitrile, 2-chloro-5-nitro, 2-chlor-5-nitrobenzonitril german, 5-nitro-2-chlorobenzonitrile, chembl52706, 2-chloro-5-nitro-benzonitrile, 2-chloro-5-nitrobenzenecarbonitrile, 4-chloro-3-cyano-1-nitrobenzene, 2-chlor-5-nitrobenzonitril, pubchem4740, 2-chloro-5-nitrobenzonitril
85504
2-chloro-5-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16588-02-6
C7H3ClN2O2
182.563
MFCD00007292
ZGILLTVEEBNDOB-UHFFFAOYSA-N
benzonitrile, 2-chloro-5-nitro, 2-chlor-5-nitrobenzonitril german, 5-nitro-2-chlorobenzonitrile, chembl52706, 2-chloro-5-nitro-benzonitrile, 2-chloro-5-nitrobenzenecarbonitrile, 4-chloro-3-cyano-1-nitrobenzene, 2-chlor-5-nitrobenzonitril, pubchem4740, 2-chloro-5-nitrobenzonitril
85504
2-chloro-5-nitrobenzonitrile
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 1818642
Formula Weight 182.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-nitrobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'