Få mere at vide
2-Chloro-5-formylbenzoic Acid, TRC
Artikelnummer. 30365485
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365485 100 mg
30357410 1 g
30362669 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365485 Leverandør Toronto Research Chemicals Leverandørnr. TRCC197676100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1206625-81-1
Molekylær formel C8H5ClO3
Molekylvægt (g/mol) 184.57
IUPAC navn 2-chloro-5-formylbenzoic acid
SMIL OC(=O)c1cc(C=O)ccc1Cl
Tekniske data
Kemisk navn eller materiale 2-Chloro-5-formylbenzoic Acid
Mængde 100 mg
InChI formel InChI=1S/C8H5ClO3/c9-7-2-1-5(4-10)3-6(7)8(11)12/h1-4H,(H,11,12)
Formel vægt 183.9927