Få mere at vide
2-Chloro-4-(trifluoromethyl)pyridine, TRC
Artikelnummer. 30355194
Skift visning
Mængde:
2.5 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355194 2.5 g
30354339 5 g
30350795 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30355194 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5974882.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81565-18-6
Molekylær formel C6H3ClF3N
Molekylvægt (g/mol) 181.54
IUPAC navn 2-chloro-4-(trifluoromethyl)pyridine
SMIL FC(F)(F)c1ccnc(Cl)c1
Tekniske data
Kemisk navn eller materiale 2-Chloro-4-(trifluoromethyl)pyridine
Mængde 2.5 g
InChI formel InChI=1S/C6H3ClF3N/c7-5-3-4(1-2-11-5)6(8,9)10/h1-3H
Formel vægt 180.9906