missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-7047394
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
175205-54-6
C7H3Cl2F3O2S
279.05
MFCD00052912
NJXDBSSSDPOAFI-UHFFFAOYSA-N
2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
2736675
2-chloro-4-(trifluoromethyl)benzenesulfonyl chloride
FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
175205-54-6
C7H3Cl2F3O2S
279.05
MFCD00052912
NJXDBSSSDPOAFI-UHFFFAOYSA-N
2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
2736675
2-chloro-4-(trifluoromethyl)benzenesulfonyl chloride
FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 39°C
Density 1.6
Boiling Point 70°C to 71°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.575
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 279.07
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'