missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7031074
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21971-21-1
C8H7ClO3
186.591
MFCD00085943
IBANGHTVBPZCHF-UHFFFAOYSA-N
2-chloro-4-methoxy-benzoic acid, 2-chloro-4-methyloxy benzoic acid, benzoic acid, 2-chloro-4-methoxy, 2-chloro-4-methoxybenzoicacid, 2-chloranissaure, pubchem14030, 2-chloro-p-anisic acid, ksc494q0b, 4-methoxy-2-chloro-benzoic acid
12648026
2-chloro-4-methoxybenzoic acid
COC1=CC(=C(C=C1)C(=O)O)Cl
This item is not returnable. View return policy
21971-21-1
C8H7ClO3
186.591
MFCD00085943
IBANGHTVBPZCHF-UHFFFAOYSA-N
2-chloro-4-methoxy-benzoic acid, 2-chloro-4-methyloxy benzoic acid, benzoic acid, 2-chloro-4-methoxy, 2-chloro-4-methoxybenzoicacid, 2-chloranissaure, pubchem14030, 2-chloro-p-anisic acid, ksc494q0b, 4-methoxy-2-chloro-benzoic acid
12648026
2-chloro-4-methoxybenzoic acid
COC1=CC(=C(C=C1)C(=O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'