missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-iodopyridine-3-carboxaldehyde, 98%

Brand:  Alfa Aesar™ H50048.03

Additional Details : CAS Number : 153034-90-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11494430

  • 1107.00 DKK / 1g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

153034-90-3
267.45
OPZCXONVRBYGHX-UHFFFAOYSA-N
10038637
C1=CN=C(C(=C1I)C=O)Cl
C6H3ClINO
MFCD03095294
2-chloro-4-iodonicotinaldehyde, 2-chloro-3-formyl-4-iodopyridine, 2-chloro-4-iodopyridine-3-carboxaldehyde, 2-chloro-4-iodo-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2-chloro-4-iodo, pubchem8084, acmc-209xa5, 2-chloro-4-iodo-3-pyridinecarbaldehyde, 3-pyridinecarboxaldehyde,2-chloro-4-iodo, 2-chloro-4-iodo-3-pyridinecarboxaldehyde
2-chloro-4-iodopyridine-3-carbaldehyde

Specifications

2-Chloro-4-iodopyridine-3-carboxaldehyde
1g
C6H3ClINO
2-chloro-4-iodonicotinaldehyde, 2-chloro-3-formyl-4-iodopyridine, 2-chloro-4-iodopyridine-3-carboxaldehyde, 2-chloro-4-iodo-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2-chloro-4-iodo, pubchem8084, acmc-209xa5, 2-chloro-4-iodo-3-pyridinecarbaldehyde, 3-pyridinecarboxaldehyde,2-chloro-4-iodo, 2-chloro-4-iodo-3-pyridinecarboxaldehyde
C1=CN=C(C(=C1I)C=O)Cl
267.45
267.45
Air and light sensitive
153034-90-3
MFCD03095294
OPZCXONVRBYGHX-UHFFFAOYSA-N
2-chloro-4-iodopyridine-3-carbaldehyde
10038637
98%
Videos
SDS Documents