Få mere at vide
2-Chloro-4-hydroxy-5-methoxybenzaldehyde, TRC
Artikelnummer. 30352185
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30352185 25 mg
30354850 50 mg
30354740 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30352185 Leverandør Toronto Research Chemicals Leverandørnr. TRCC61162025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18268-76-3
Molekylær formel C8 H7 Cl O3
Molekylvægt (g/mol) 186.59
Synonym 6-Chlorovanillin, Benzaldehyde, 2-chloro-4-hydroxy-5-methoxy-, Vanillin, 6-chloro- (8CI), 2-Chloro-4-hydroxy-5-methoxybenzaldehyde, 6-Chlorovanillin, 6-Monochlorovanillin, NSC 95796
IUPAC navn 2-chloro-4-hydroxy-5-methoxybenzaldehyde
SMIL COc1cc(C=O)c(Cl)cc1O
Tekniske data
Kemisk navn eller materiale 6-Chlorovanillin
Mængde 25 mg
InChI formel InChI=1S/C8H7ClO3/c1-12-8-2-5(4-10)6(9)3-7(8)11/h2-4,11H,1H3
Formel vægt 186.0084