Få mere at vide
2-Chloro-4-(3-piperidinyloxy)benzonitrile, 97%, Thermo Scientific™
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Chloro-4-(3-piperidinyloxy)benzonitrile |
| Mængde | 250 mg |
| Opløselighedsinformation | Insoluble in water. |
| Formel vægt | 236.7 |
| Procent renhed | 97% |