Få mere at vide
2-Chloro-3-methylquinoline, 97%, Thermo Scientific™
Artikelnummer. 11345569
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11345569 5 g 5g
11335569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11345569 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H50492.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Chloro-3-methylquinoline is used as organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57876-69-4
Molekylær formel C10H8ClN
Molekylvægt (g/mol) 177.631
MDL nummer MFCD00100265
InChI nøgle UMYPPRQNLXTIEQ-UHFFFAOYSA-N
Synonym 2-chloro-3-methyl-quinoline, quinoline, 2-chloro-3-methyl, zlchem 105, pubchem12431, acmc-1ayrn, methyl 2-chloroquinoline, 2-chloro-3-methylquinolin, 3-methyl-2-chloroquinoline, 2-cloro-3-methyl-quinoline, 2-chloro-3-methyl quinoline
PubChem CID 339082
IUPAC navn 2-chloro-3-methylquinoline
SMIL CC1=CC2=CC=CC=C2N=C1Cl
Tekniske data
Kemisk navn eller materiale 2-Chloro-3-methylquinoline
Smeltepunkt 82°C to 84°C
Mængde 5 g
Opløselighedsinformation Insoluble in water.
Formel vægt 177.63
Procent renhed 97%