missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Chloro-3-hydroxybenzoic Acid, TRC

Artikelnummer. 30350211
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350211 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30350211 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58579550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Chloro-3-hydroxybenzoic Acid
CAS 51786-10-8
Molekylær formel C7H5O3Cl
Mængde 50 mg
InChI formel InChI=1S/C7H5ClO3/c8-6-4(7(10)11)2-1-3-5(6)9/h1-3,9H,(H,10,11)
SMIL C1=CC(=C(C(=C1)O)Cl)C(=O)O
IUPAC navn 2-chloro-3-hydroxybenzoic acid
Molekylvægt (g/mol) 172.57
Formel vægt 171.9927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.