Få mere at vide
2-Chloro-3-fluoro-4-(trifluoromethyl)benzoic acid, TRC
Artikelnummer. 30364147
Skift visning
Mængde:
500 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364147 500 mg
30351102 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364147 Leverandør Toronto Research Chemicals Leverandørnr. TRCC598471500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro-3-fluoro-4-(trifluoromethyl)benzoic acid
CAS 1805457-37-7
Molekylær formel C8H3ClF4O2
Mængde 500 mg
InChI formel InChI=1S/C8H3ClF4O2/c9-5-3(7(14)15)1-2-4(6(5)10)8(11,12)13/h1-2H,(H,14,15)
SMIL OC(=O)c1ccc(c(F)c1Cl)C(F)(F)F
IUPAC navn 2-chloro-3-fluoro-4-(trifluoromethyl)benzoic acid
Molekylvægt (g/mol) 242.55
Formel vægt 241.9758