Learn More
2-Chloro-3,4-dimethoxypyridine, 98%, BLD Pharm™
Product Code. 31030649
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31030649 100 mg -
31027275 1 g -
31016153 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31030649 Supplier BLD Pharm Supplier No. BD00802296100mg
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 101664-59-9
Molecular Formula C7H8ClNO2
MDL Number MFCD03265283
Quantity 100 mg
SMILES COC1=C(OC)C(Cl)=NC=C1
Molecular Weight (g/mol) 173.60
Percent Purity 98%
Chemical Name or Material 2-Chloro-3,4-dimethoxypyridine