Få mere at vide
2-Chloro-3,4-dihydroxybenzoic acid, TRC
Artikelnummer. 30367760
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367760 100 mg
30360750 10 mg
30352356 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367760 Leverandør Toronto Research Chemicals Leverandørnr. TRCC598093100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 87932-50-1
Molekylær formel C7H5ClO4
Molekylvægt (g/mol) 188.56
IUPAC navn 2-chloro-3,4-dihydroxybenzoic acid
SMIL OC(=O)c1ccc(O)c(O)c1Cl
Tekniske data
Kemisk navn eller materiale 2-Chloro-3,4-dihydroxybenzoic acid
Mængde 100 mg
InChI formel InChI=1S/C7H5ClO4/c8-5-3(7(11)12)1-2-4(9)6(5)10/h1-2,9-10H,(H,11,12)
Formel vægt 187.9876